CaptureSelect™ IgG1 (Hu) Affinity Matrix

Catalog number:  191303050

Thermo Scientific™  Related applications: Antibody Purification | Chromatography | Protein Purification & Isolation

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

CaptureSelect™ IgG1 (Hu) Affinity Matrix can be used for the purification and isolation of IgG1, without cross-reactivity to other immunoglobulins, like other human IgG subclasses, IgA, and IgM, from complex sources like plasma, serum, and cell culture supernatants.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Column Type: Bulk Resin
Format: Bulk Media
Immobilized Phase: Anti IgG1
Product Line: CaptureSelect™
Product Size: 50 mL

Contents & storage

Store at 2–8°C.

Documents

Manuals & protocols