Search
| Número de catálogo | Cantidad |
|---|---|
| 191304005 | 5 mL |
| 191304010 | 10 mL |
| 191304050 | 50 mL |
CaptureSelect IgG3 (Hu) Affinity Matrix can be used for the purification and isolation of IgG3, without cross-reactivity to other immunoglobulins, like other human IgG subclasses, IgA, and IgM, from complex sources like plasma, serum, and cell culture supernatants.