CaptureSelect™ Transferrin Affinity Matrix
CaptureSelect™ Transferrin Affinity Matrix
Thermo Scientific™
CaptureSelect™ Transferrin Affinity Matrix
This is one of a group of Thermo Scientific™ CaptureSelect™ protein purification affinity resins developed for the simple, single-step purification of a variety human plasma and serum proteins.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
19130605050 mL
1913060055 mL
19130601010 mL
Catalog number 191306050
Price (USD)
10,930.00
Each
Add to cart
Quantity:
50 mL
Price (USD)
10,930.00
Each
Add to cart
This is one of a group of CaptureSelect protein purification affinity resins developed for the simple, single-step purification of a variety human plasma and serum proteins. This unique selection of affinity purification resins can purify proteins or other non-antibody biomolecules from either recombinant or native sources with a high degree of affinity and specificity, and are immobilized on a high capacity agarose support.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Quantity50 mL
Stationary PhaseAnti-transferrin
FormLiquid Suspension
Product LineCaptureSelect
TypeAffinity Matrix
Unit SizeEach
Contents & Storage
Store at 2°C to 8°C.