CaptureSelect™ C-tag Affinity Matrix

Catalog number: 191307010

 Related applications: Chromatography | Protein Purification & Isolation

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The CaptureSelect™ C-tag Affinity Matrix combines unique selectivity for a small 4-amino acid peptide tag (E-P-E-A, glutamic acid-proline-glutamic acid-alanine) with the benefits of a robust and high quality affinity matrix. The CaptureSelect™ C-tag Affinity Matrix purifies C-terminal tagged proteins with high affinity and selectivity, even in the presence of urea and guanidine HCl, from complex mixtures like cytoplasm or periplasmatic fractions in a one-step process. Mild elution conditions at neutral pH can be applied using magnesium chloride or propylene glycol, which ensures high activity recoveries of pH-sensitive target proteins. The affinity resin recognizes the E-P-E-A tag sequence when fused either directly to the C-terminus of a protein or through linkers between the C-terminus and the E-P-E-A tag.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Column Type: Bulk Resin
Format: Bulk Media
Immobilized Phase: Anti C-tag
Product Line: CaptureSelect™
Product Size: 10 mL

Contents & storage

Store at 2–8°C.

Documents

Manuals & protocols