CaptureSelect™ LC-lambda (ung) Affinity Matrix

Catalog number: 191314050

 Related applications: Antibody Purification | Chromatography | Protein Purification & Isolation

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

This is one of a group of CaptureSelect™ Antibody Toolbox™ affinity resins designed for the simple, single-step purification of all antibody subtypes and fragments. Unlike the common bacterial coat protein matrices (e.g., Protein A, G, and L), the Antibody Toolbox resins were specifically designed to purify any antibody format with high affinity and specificity. These unique affinity ligands are immobilized on a high capacity agarose support for lower-pressure antibody purification applications.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Column Type: Bulk Resin
Format: Bulk Media
Immobilized Phase: Anti LC-lambda
Product Line: CaptureSelect™
Product Size: 50 mL

Contents & storage

Store at 2–8°C.

Documents

Manuals & protocols