CaptureSelect™ LC-kappa (mur) Affinity Matrix
Thermo Scientific™

CaptureSelect™ LC-kappa (mur) Affinity Matrix

Thermo Scientific™ CaptureSelect™ LC-kappa (mur) Affinity Matrix enables purification of murine (mouse, rat, and guinea pig) immunoglobulins and Fab fragments without cross reactivity towards bovine immunoglobulins.
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoCantidad
19131505050 mL
1913150055 mL
19131501010 mL
Número de catálogo 191315050
Precio (CLP)
-
Cantidad:
50 mL

CaptureSelect LC-kappa (mur) Affinity Matrix enables purification of murine (mouse, rat, and guinea pig) immunoglobulins and Fab fragments without cross reactivity towards bovine immunoglobulins. The ligand is directed towards a unique domain on the constant part of the kappa light chain of immunoglobulins, which makes CaptureSelect LC-kappa (mur) Affinity Matrix a unique resin in terms of specificity.

This matrix can be used for the purification and isolation of kappa light chain-containing murine immunoglobulins and Fab fragments from complex sources like plasma, serum, and cell culture supernatants in a single step. Due to its non-reactivity with bovine Ig, FCS can be applied in growth media without disturbing purification.

Para uso exclusivo en investigación. No apto para uso en procedimientos diagnósticos.
Especificaciones
Cantidad50 mL
Fase estacionariaAnti-LC-kappa (mur)
FormularioSuspensión líquida
Tamaño de partículas35 μm
Línea de productosCaptureSelect
TipoAffinity Matrix
Unit SizeEach
Contenido y almacenamiento
Store at 2°C to 8°C.