Fluorescein diacetate, 97%, pure
Fluorescein diacetate, 97%, pure
Fluorescein diacetate, 97%, pure
Thermo Scientific Chemicals

Fluorescein diacetate, 97%, pure

CAS: 596-09-8 | C24H16O7 | 416.39 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 191661000
Price (USD)
725.00
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
725.00
Each
Chemical Identifiers
CAS596-09-8
IUPAC Name6'-(acetyloxy)-3-oxo-3H-spiro[2-benzofuran-1,9'-xanthen]-3'-yl acetate
Molecular FormulaC24H16O7
InChI KeyCHADEQDQBURGHL-UHFFFAOYSA-N
SMILESCC(=O)OC1=CC=C2C(OC3=CC(OC(C)=O)=CC=C3C22OC(=O)C3=CC=CC=C23)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Light yellow
Melting point200°C to 205°C
HPLC>=96.0 %
Infrared spectrumConforms
Sulfated ash=<0.1 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only