Fluorescamine, pure
Fluorescamine, pure
Fluorescamine, pure
Thermo Scientific Chemicals

Fluorescamine, pure

CAS: 38183-12-9 | C17H10O4 | 278.26 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mg
100 mg
500 mg
2.5 g
Packaging:
Glass bottle
Catalog number 191670250
Price (USD)
47.60
Each
Quantity:
25 mg
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
47.60
Each
Chemical Identifiers
CAS38183-12-9
IUPAC Name4'-phenyl-3H,3'H-spiro[2-benzofuran-1,2'-furan]-3,3'-dione
Molecular FormulaC17H10O4
InChI KeyZFKJVJIDPQDDFY-UHFFFAOYNA-N
SMILESO=C1OC2(OC=C(C2=O)C2=CC=CC=C2)C2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Specific extinction E (1%/1cm)>=900 (235 nm, ether)
Infrared spectrumConforms
Suitabilityfluorescence ind. for prim. amines and amino acids
Appearance (Form)Powder
Melting point155°C to 157°C
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only