Ruthenium Red tetrahydrate, pure
Ruthenium Red tetrahydrate, pure
Ruthenium Red tetrahydrate, pure
Thermo Scientific Chemicals

Ruthenium Red tetrahydrate, pure

CAS: 12790-48-6 | Cl6H36N14O6Ru3-14 | 844.29 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 191730010
Price (TWD)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS12790-48-6
IUPAC Namebis(λ⁵-ruthenium(5+)) tetrahydrate ruthenium tetradecaazanide hexachloride dioxidandiide
Molecular FormulaCl6H36N14O6Ru3
InChI KeyIMVKVBRYNZXYAP-UHFFFAOYSA-H
SMILES[NH2-].[NH2-].[NH2-].[NH2-].[NH2-].[NH2-].[NH2-].[NH2-].[NH2-].[NH2-].[NH2-].[NH2-].[NH2-].[NH2-].O.O.O.O.[O--].[O--].[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Ru].[Ru+5].[Ru+5]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Dark red to dark brown to black
Appearance (Form)Powder
SolubilityClear (0.1% aq. solution)
UV Lambda max529 to 539 nm
Specific absorbance A (1%/1cm)>=80 (1% aq. solution in a 1cm cell v/s H2O)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only