Dextrose, anhydrous, 97.5-102.0% (by anhydrous basis), USP, GMP, J.T.Baker™
Dextrose, anhydrous, 97.5-102.0% (by anhydrous basis), USP, GMP, J.T.Baker™
Dextrose, anhydrous, 97.5-102.0% (by anhydrous basis), USP, GMP, J.T.Baker™
Avantor™

Dextrose, anhydrous, 97.5-102.0% (by anhydrous basis), USP, GMP, J.T.Baker™

This product is GMP Manufactured and meets USP specifications.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityPackaging
1919-09100 lb.Fiber Drum
1919-01500 gPoly Bottle, Tamper-evident Cap
1919-25200 lb.Polyethylene Drum, Open Head
1919-052.5 kgPoly Bottle, Tamper-evident Cap
1919-0712 kgPolyethylene Pail, Screw Top
1919-08200 lb.Fiber Drum
Catalog number 1919-09
Price (CLP)
-
Quantity:
100 lb.
Packaging:
Fiber Drum
This product is GMP Manufactured and meets USP specifications.
This product is GMP Manufactured and meets USP specifications.
Specifications
CAS50-99-7
ColorWhite
IUPAC Name6-(hydroxymethyl)oxane-2,3,4,5-tetrol
InChI KeyWQZGKKKJIJFFOK-UHFFFAOYNA-N
Melting Point146°C
Molecular FormulaC6H12O6
PackagingFiber Drum
SMILESOCC1OC(O)C(O)C(O)C1O
GradeUSP
Molecular Weight (g/mol)180.16
Percent Purity97.5 to 102%
Physical FormSolid
Quantity100 lb.
Unit SizeEach