Dextrose, anhydrous, 97.5-102.0% (by anhydrous basis), USP, Multi-Compendial, GMP, J.T.Baker™
Avantor™

Dextrose, anhydrous, 97.5-102.0% (by anhydrous basis), USP, Multi-Compendial, GMP, J.T.Baker™

This product is GMP Manufactured and meets USP specifications.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
1920-052.5 kg
1920-09100 lb.
1920-0712 kg
1920-06200 lb.
1920-20100 kg
Catalog number 1920-05
Price (MXN)
-
Quantity:
2.5 kg
This product is GMP Manufactured and meets USP specifications.
This product is GMP Manufactured and meets USP specifications.
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Specifications
ColorWhite
Melting Point146°C
PackagingPoly bottle, tamper-evident cap
GradeUSP
Molecular Weight (g/mol)180.16
Percent Purity97.5 to 102%
Physical FormSolid
Quantity2.5 kg
Unit SizeEach