Dextrose, anhydrous, 97.5-102.0% (by anhydrous basis), USP, Multi-Compendial, GMP, J.T.Baker™
Avantor™

Dextrose, anhydrous, 97.5-102.0% (by anhydrous basis), USP, Multi-Compendial, GMP, J.T.Baker™

This product is GMP Manufactured and meets USP specifications.Más información
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoCantidad
1920-0712 kg
1920-09100 lb
1920-052,5 kg
1920-06200 lb
1920-20100 kg
Número de catálogo 1920-07
Precio (MXN)
-
Cantidad:
12 kg
This product is GMP Manufactured and meets USP specifications.
Especificaciones
CAS50-99-7
ColorBlanco
Punto de fusión146°C
EnvasePolyethylene pail, screw top
GradoUSP
Peso molecular (g/mol)180.16
Porcentaje de pureza97.5 to 102%
Forma físicaSolid
Cantidad12 kg
Unit SizeEach