Dextrose, anhydrous, 97.5-102.0% (by anhydrous basis), USP, Multi-Compendial, GMP, J.T.Baker™
Avantor™

Dextrose, anhydrous, 97.5-102.0% (by anhydrous basis), USP, Multi-Compendial, GMP, J.T.Baker™

This product is GMP Manufactured and meets USP specifications.
Have Questions?
Alterar visualizaçãobuttonViewtableView
Número do catálogoQuantity
1920-09100 lb.
1920-052.5 kg
1920-0712 kg
1920-06200 lb.
1920-20100 kg
Número do catálogo 1920-09
Preço (BRL)
-
Quantity:
100 lb.
This product is GMP Manufactured and meets USP specifications.
This product is GMP Manufactured and meets USP specifications.
Especificações
CAS50-99-7
ColorWhite
Melting Point146°C
PackagingPoly drum
GradeUSP
Molecular Weight (g/mol)180.16
Percent Purity97.5 to 102%
Physical FormSolid
Quantity100 lb.
Unit SizeEach