Cesium carbonate, 99.5%, for analysis
Cesium carbonate, 99.5%, for analysis
Cesium carbonate, 99.5%, for analysis
Thermo Scientific Chemicals

Cesium carbonate, 99.5%, for analysis

CAS: 534-17-8 | CCs2O3 | 325.82 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Catalog number 192045000
Price (USD)
527.65
Special offer
Online exclusive
Ends: 30-Jun-2026
621.00
Save 93.35 (15%)
Each
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
527.65
Special offer
Online exclusive
Ends: 30-Jun-2026
621.00
Save 93.35 (15%)
Each
Chemical Identifiers
CAS534-17-8
IUPAC Namedicaesium(1+) carbonate
Molecular FormulaCCs2O3
InChI KeyFJDQFPXHSGXQBY-UHFFFAOYSA-L
SMILES[Cs+].[Cs+].[O-]C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Barium (Ba)=<100 ppm
Appearance (Form)Crystalline powder
Sodium (Na)=<300 ppm
Sulfate (SO4)=<500 ppm
Appearance (Color)White
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only