Transferrin, Bovine APO UL IGG
Gibco™
Transferrin, Bovine APO UL IGG
Iron-free, low IgG bovine transferrin for cell culture. It is chromatographically purifed, has an iron content ≤50 μg/g protein and an IgG content ≤50ppm. Lyophilized powder.
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoCantidad
1921004IJ5 g
1921004IC10 g
Número de catálogo 1921004IJ
Precio (CLP)
-
Cantidad:
5 g
Iron-free, low-IgG bovine transferrin for cell culture. It is chromatographically purified and has an iron content ≤50 μg/g protein and an IgG content ≤50 ppm. Lyophilized powder.
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Especificaciones
Tipo de célulaMammalian Cells
Tipo de productoTransferrin
Cantidad5 g
FormularioSolid
Unit SizeEach