Poly(propylene glycol), average M.W. 2.000
Poly(propylene glycol), average M.W. 2.000
Poly(propylene glycol), average M.W. 2.000
Acros Organics

Poly(propylene glycol), average M.W. 2.000

CAS: 25322-69-4 | (C3H6O)n | 134.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
500 g
Packaging:
Glass bottle
Catalog number 192180025
also known as 19218
HSN 39072090
Price (INR)
33,975.00
Each
Quantity:
2.5 kg
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
33,975.00
Each
Chemical Identifiers
CAS25322-69-4
Molecular Formula(C3H6O)n
InChI KeyDNIAPMSPPWPWGF-UHFFFAOYNA-N
SMILES[H]OC(C)CO
MDL NumberMFCD00084446
SpecificationsSpecification SheetSpecification Sheet
Color scale=<50 APHA
Refractive index1.4490 to 1.4530 (20°C, 589 nm)
Appearance (Form)Clear viscous liquid
Acid value=<0.1 mg KOH/g
Viscosity305 to 335 cSt (at 25°C)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only