This product ships via ground from US northeast.
Poly(propylene glycol), average M.W. 4.000
Poly(propylene glycol), average M.W. 4.000
Poly(propylene glycol), average M.W. 4.000
Thermo Scientific Chemicals

Poly(propylene glycol), average M.W. 4.000

CAS: 25322-69-4 | (C3H6O)n | 134.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
500 g
Packaging:
Glass bottle
Catalog number 192205000
Price (USD)
69.60
Each
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
69.60
Each
Chemical Identifiers
CAS25322-69-4
Molecular Formula(C3H6O)n
InChI KeyDNIAPMSPPWPWGF-UHFFFAOYNA-N
SMILES[H]OC(C)CO
MDL NumberMFCD00084446
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear light yellow to light green
Infrared spectrumConforms
Appearance (Form)Viscous liquid
Refractive index1.4500 to 1.4520 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only