Poly(ethylene glycol), average M.W. 300
Poly(ethylene glycol), average M.W. 300
Poly(ethylene glycol), average M.W. 300
Thermo Scientific Chemicals

Poly(ethylene glycol), average M.W. 300

CAS: 25322-68-3 | (C2H4O)n | 62.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
10 L
Packaging:
View all
Glass bottle
Catalog number 192220010
Price (USD)
90.50
Each
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
90.50
Each
Chemical Identifiers
CAS25322-68-3
Molecular Formula(C2H4O)n
InChI KeyLYCAIKOWRPUZTN-UHFFFAOYSA-N
SMILES[H]OCCO
SpecificationsSpecification SheetSpecification Sheet
Color scale=<50 APHA
Infrared spectrumConforms
Hydroxyl value356 to 394 mg KOH/g
Water=<0.5 % (K.F.)
Appearance (Form)Clear liquid
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only