Poly(ethylene glycol), average M.W. 6000
Poly(ethylene glycol), average M.W. 6000
Poly(ethylene glycol), average M.W. 6000
Acros Organics

Poly(ethylene glycol), average M.W. 6000

CAS: 25322-68-3 | (C2H4O)n | 62.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 kg
1 kg
Packaging:
View all
Plastic drum
Catalog number 192280051
HSN 39072090
Price (INR)
30,442.00
Each
Quantity:
5 kg
Packaging:
Plastic drum
Request bulk or custom format
Price (INR)
30,442.00
Each
Chemical Identifiers
CAS25322-68-3
Molecular Formula(C2H4O)n
InChI KeyLYCAIKOWRPUZTN-UHFFFAOYSA-N
SMILES[H]OCCO
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Hydroxyl value17 to 20 mg KOH/g
Water=<0.5 % (K.F.)
Appearance (Form)Powder or flakes
Appearance (Color)White
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only