Thermo Scientific Chemicals

Poly(ethylene glycol methyleether), average M.W. 350, Thermo Scientific Chemicals

Catalog number: 192300010
1 kg, Glass bottle, Each
Thermo Scientific Chemicals

Poly(ethylene glycol methyleether), average M.W. 350, Thermo Scientific Chemicals

Catalog number: 192300010
1 kg, Glass bottle, Each
Quantity
Packaging

Chemical Identifiers

CAS
9004-74-4
Molecular Formula
(C2H4O)nCH4O
InChI Key
XNWFRZJHXBZDAG-UHFFFAOYSA-N
SMILES
COCCO
MDL Number
MFCD00084416
Infrared spectrum
Conforms
Color scale
=<100 APHA
Appearance (Form)
Clear liquid
Hydroxyl value
150 to 172 mg KOH/g
Water
=<0.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text