Aluminum sulfate, 99.999%, (trace metal basis), extra pure
Thermo Scientific Chemicals

Aluminum sulfate, 99.999%, (trace metal basis), extra pure

CAS: 10043-01-3 | Al2O12S3 | 342.13 g/mol
数量:
5 g
25 g
100 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 192431000
価格(JPY)
-
数量:
100 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS10043-01-3
IUPAC Namealuminium(3+) potassium disulfate
Molecular FormulaAlKO8S2
InChI KeyGRLPQNLYRHEGIJ-UHFFFAOYSA-J
SMILES[Al+3].[K+].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
さらに表示
Titration Complexometric>=15 % (Al) Typical
Appearance (Form)Powder and/or chunks
Total trace metal impurities=<0.001 %
Appearance (Color)White
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysis
RUO – Research Use Only