Europium(III) oxide, 99.99%, (trace metal basis)
Europium(III) oxide, 99.99%, (trace metal basis)
Europium(III) oxide, 99.99%, (trace metal basis)
Thermo Scientific Chemicals

Europium(III) oxide, 99.99%, (trace metal basis)

CAS: 1308-96-9 | Eu2O3 | 351.93 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Catalog number 193020250
Price (EUR)
345,00
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
345,00
Each
Chemical Identifiers
CAS1308-96-9
IUPAC Namedieuropium(3+) trioxidandiide
Molecular FormulaEu2O3
InChI KeyAEBZCFFCDTZXHP-UHFFFAOYSA-N
SMILES[O--].[O--].[O--].[Eu+3].[Eu+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light pink
Appearance (Form)Powder
Total conc. of imp.as REO=<0.1 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for ultra trace analysis
RUO – Research Use Only