Indium(III) chloride, 99.995%, (trace metal basis)
Indium(III) chloride, 99.995%, (trace metal basis)
Indium(III) chloride, 99.995%, (trace metal basis)
Thermo Scientific Chemicals

Indium(III) chloride, 99.995%, (trace metal basis)

CAS: 10025-82-8 | Cl3In | 221.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
Catalog number 193192500
Price (USD)
-
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS10025-82-8
IUPAC Nameindium(3+) trichloride
Molecular FormulaCl3In
InChI KeyPSCMQHVBLHHWTO-UHFFFAOYSA-K
SMILES[Cl-].[Cl-].[Cl-].[In+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Total trace metal impurities=<0.005 %
Appearance (Color)White
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysis
RUO – Research Use Only