Iron(III) oxide, 99.999%, (trace metal basis), -100 mesh
Iron(III) oxide, 99.999%, (trace metal basis), -100 mesh
Iron(III) oxide, 99.999%, (trace metal basis), -100 mesh
Thermo Scientific Chemicals

Iron(III) oxide, 99.999%, (trace metal basis), -100 mesh

CAS: 1309-37-1 | Fe2O3 | 159.69 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Packaging:
View all
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 193260500
Price (EUR)
-
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS1309-37-1
IUPAC Nametrioxodiiron
Molecular FormulaFe2O3
InChI KeyJEIPFZHSYJVQDO-UHFFFAOYSA-N
SMILES[Fe][Fe](=O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Red-brown to rust brown
Appearance (Form)Powder
Total trace metal impurities=<0.001 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for ultra trace analysis
RUO – Research Use Only