Lanthanum(III) nitrate hexahydrate, 99.995%, (trace metal basis)
Lanthanum(III) nitrate hexahydrate, 99.995%, (trace metal basis)
Lanthanum(III) nitrate hexahydrate, 99.995%, (trace metal basis)
Thermo Scientific Chemicals

Lanthanum(III) nitrate hexahydrate, 99.995%, (trace metal basis)

CAS: 10277-43-7 | H12LaN3O15 | 433.01 g/mol
数量:
100 g
500 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 193281000
価格(JPY)
-
数量:
100 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS10277-43-7
IUPAC Namelanthanum(3+) hexahydrate trinitrate
Molecular FormulaH12LaN3O15
InChI KeyGJKFIJKSBFYMQK-UHFFFAOYSA-N
SMILESO.O.O.O.O.O.[La+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
さらに表示
Appearance (Form)Crystals and/or chunks
Lanthanum (La)31.0 to 35.0 %
Total trace metal impurities=<0.005 %
Appearance (Color)White
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysis
RUO – Research Use Only