Lead(II) nitrate, 99.999%, (trace metal basis)
Lead(II) nitrate, 99.999%, (trace metal basis)
Lead(II) nitrate, 99.999%, (trace metal basis)
Thermo Scientific Chemicals

Lead(II) nitrate, 99.999%, (trace metal basis)

CAS: 10099-74-8 | N2O6Pb | 331.20 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Packaging:
Glass bottle
Catalog number 193320100
Price (USD)
58.65
Special offer
Online exclusive
Ends: 28-Dec-2026
69.30
Save 10.65
Each
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
58.65
Special offer
Online exclusive
Ends: 28-Dec-2026
69.30
Save 10.65
Each
Chemical Identifiers
CAS10099-74-8
IUPAC Nameλ²-lead(2+) dinitrate
Molecular FormulaN2O6Pb
InChI KeyRLJMLMKIBZAXJO-UHFFFAOYSA-N
SMILES[Pb++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total trace metal impurities=<0.001 %
Appearance (Form)Crystalline solid
Appearance (Color)White
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysis
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only