Lithium carbonate, 99.999%, (trace metal basis), extra pure
Lithium carbonate, 99.999%, (trace metal basis), extra pure
Lithium carbonate, 99.999%, (trace metal basis), extra pure
Thermo Scientific Chemicals

Lithium carbonate, 99.999%, (trace metal basis), extra pure

CAS: 554-13-2 | CLi2O3 | 73.89 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
100 g
Packaging:
Glass bottle
Catalog number 193360100
Price (USD)
52.90
Each
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
52.90
Each
Chemical Identifiers
CAS554-13-2
IUPAC Namedilithium(1+) carbonate
Molecular FormulaCLi2O3
InChI KeyXGZVUEUWXADBQD-UHFFFAOYSA-L
SMILES[Li+].[Li+].[O-]C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Powder
Total trace metal impurities=<0.001 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysis
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only