Nitrato de manganeso(II) tetrahidrato, para análisis, Thermo Scientific Chemicals
Nitrato de manganeso(II) tetrahidrato, para análisis, Thermo Scientific Chemicals
Nitrato de manganeso(II) tetrahidrato, para análisis, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Nitrato de manganeso(II) tetrahidrato, para análisis, Thermo Scientific Chemicals

CAS: 20694-39-7 | MnN2O6·4H2O | 251.01 g/mol
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
250 g
1 kg
Envase:
Frasco de plástico
Número de catálogo 193462500
Precio (USD)
-
Cantidad:
250 g
Envase:
Frasco de plástico
Pedido a granel o personalizado
Identificadores químicos
CAS20694-39-7
IUPAC Namemanganese(2+) tetrahydrate dinitrate
Molecular FormulaH8MnN2O10
InChI KeyALIMWUQMDCBYFM-UHFFFAOYSA-N
SMILESO.O.O.O.[Mn++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
Ver más
EspecificacionesSpecification SheetHoja de especificaciones
Sulfate (SO4)=<0.005 %
Cadmium (Cd)=<0.0005 %
Potassium (K)=<0.01 %
Sodium (Na)=<0.05 %
Lead (Pb)=<0.001 %
Ver más
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only