Nickel(II) chloride hexahydrate, 99.999%, (trace metal basis)
Nickel(II) chloride hexahydrate, 99.999%, (trace metal basis)
Nickel(II) chloride hexahydrate, 99.999%, (trace metal basis)
Thermo Scientific Chemicals

Nickel(II) chloride hexahydrate, 99.999%, (trace metal basis)

CAS: 7791-20-0 | Cl2Ni·6H2O | 237.71 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 193570250
Price (USD)
238.65
Special offer
Online exclusive
Ends: 28-Dec-2026
280.00
Save 41.35
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
238.65
Special offer
Online exclusive
Ends: 28-Dec-2026
280.00
Save 41.35
Each
Chemical Identifiers
CAS7791-20-0
IUPAC Namenickel(2+) hexahydrate dichloride
Molecular FormulaCl2H12NiO6
InChI KeyLAIZPRYFQUWUBN-UHFFFAOYSA-L
SMILESO.O.O.O.O.O.[Cl-].[Cl-].[Ni++]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals and/or chunks
Nickel (Ni)24 to 26 %
Total trace metal impurities=<0.001 %
Appearance (Color)Green
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysis
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only