Thermo Scientific Chemicals

Hexafluorosilicato de amonio, 99,999 %, (base de trazas metálicas), Thermo Scientific Chemicals

Número de catálogo: 194000500
50 g, Frasco de vidrio, Each
Thermo Scientific Chemicals

Hexafluorosilicato de amonio, 99,999 %, (base de trazas metálicas), Thermo Scientific Chemicals

Número de catálogo: 194000500
50 g, Frasco de vidrio, Each
Cantidad
Envase
Número de catálogo: 194000500
Precio (EUR)
Cantidad
-

Identificadores químicos

CAS
16919-19-0
IUPAC Name
diammonium hexafluorosilanediuide
Molecular Formula
F6H8N2Si
InChI Key
ITHIMUMYFVCXSL-UHFFFAOYSA-P
SMILES
[NH4+].[NH4+].F[Si--](F)(F)(F)(F)F
Total trace metal impurities
=<10 ppm
Appearance (Color)
White
Appearance (Form)
Crystalline powder and/or chunks

Descripción

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Para análisis de ultratrazas
RUO – Research Use Only

Figuras

Documentos y descargas

Certificados

    Preguntas frecuentes

    Citas y referencias

    Search citations by name, author, journal title or abstract text

    Seguridad y manipulación


    Classification of the substance or mixture
    CLP classification - Regulation(EC) No 1272/2008
    Acute oral toxicity
    Category 3
    Acute dermal toxicity
    Category 3
    Acute Inhalation Toxicity - Dusts and Mists
    Category 3
    Label Elements
    Signal Word

    Danger

    Hazard Statements

    H301 + H311 + H331 - Toxic if swallowed, in contact with skin or if inhaled

    Precautionary Statements

    P280 - Wear protective gloves/protective clothing

    P301 + P310 - IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician

    P302 + P352 - IF ON SKIN: Wash with plenty of soap and water

    P304 + P340 - IF INHALED: Remove person to fresh air and keep comfortable for breathing

    P311 - Call a POISON CENTER or doctor/physician