Ammonium hexafluorosilicate, 99.999%, (trace metal basis)
Ammonium hexafluorosilicate, 99.999%, (trace metal basis)
Ammonium hexafluorosilicate, 99.999%, (trace metal basis)
Thermo Scientific Chemicals

Ammonium hexafluorosilicate, 99.999%, (trace metal basis)

CAS: 16919-19-0 | H8F6N2Si | 178.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
Catalog number 194002500
Price (TWD)
-
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS16919-19-0
IUPAC Namediammonium hexafluorosilanediuide
Molecular FormulaF6H8N2Si
InChI KeyITHIMUMYFVCXSL-UHFFFAOYSA-P
SMILES[NH4+].[NH4+].F[Si--](F)(F)(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Total trace metal impurities=<10 ppm
Appearance (Color)White
Appearance (Form)Crystalline powder and/or chunks
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for ultra trace analysis
RUO – Research Use Only