Strontium chloride hexahydrate, 99%, extra pure
Strontium chloride hexahydrate, 99%, extra pure
Strontium chloride hexahydrate, 99%, extra pure
Acros Organics

Strontium chloride hexahydrate, 99%, extra pure

CAS: 10025-70-4 | Cl2H12O6Sr | 266.61 g/mol
Have Questions?
Catalog NumberQuantityPackaging
194122500250 gPlastic bottle
Catalog number 194122500
HSN 28273990
Price (INR)
8,200.00
Each
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (INR)
8,200.00
Each
Chemical Identifiers
CAS10025-70-4
IUPAC Namestrontium(2+) hexahydrate dichloride
Molecular FormulaCl2H12O6Sr
InChI KeyAMGRXJSJSONEEG-UHFFFAOYSA-L
SMILESO.O.O.O.O.O.[Cl-].[Cl-].[Sr++]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Crystals
Titration Complexometric>=98.5 %
Barium (Ba)=<0.1 %
Calcium (Ca)=<0.1 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only