Ammonium sulfate, 99.9995%, (trace metal basis)
Ammonium sulfate, 99.9995%, (trace metal basis)
Ammonium sulfate, 99.9995%, (trace metal basis)
Thermo Scientific Chemicals

Ammonium sulfate, 99.9995%, (trace metal basis)

CAS: 7783-20-2 | H8N2O4S | 132.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Packaging:
Glass bottle
Catalog number 194150500
Price (USD)
96.65
Online exclusive
107.00
Save 10.35
Each
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
96.65
Online exclusive
107.00
Save 10.35
Each
Chemical Identifiers
CAS7783-20-2
IUPAC Namesulfuric acid diamine
Molecular FormulaH8N2O4S
InChI KeyBFNBIHQBYMNNAN-UHFFFAOYSA-N
SMILESN.N.OS(O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Crystalline powder or crystals and/or chunks
Gravimetry23.7 to 24.9 % (S)
Total trace metal impurities=<5 ppm
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysis
RUO – Research Use Only