Thermo Scientific™

CaptureSelect™ IgA Affinity Matrix

Catalog number:  19428801L

 Related applications: Antibody Purification | Protein Purification & Isolation

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The CaptureSelect™ IgA Affinity Matrix has specifically been designed for the purification of human IgA from recombinant and plasma sources.

Features of this affinity matrix include:

• Platform for all human IgA, binding to IgA1, IgA2 , dimeric IgA, and S-IgA (secretory IgA)
• Excellent scalability
• Non-animal-derived

Free of Animal Components
Our CaptureSelect™ products are affinity ligands created by a proprietary technology based on Camelid-derived single domain antibody fragments. The ligand is a 13 kDa single domain fragment comprising the 3 CDRs that form the antigen binding domain, efficiently produced by the yeast Saccharomyces cerevisiae in a production process free of animal components.

Main Characteristics:

Matrix: agarose-based, aldehyde-activated
Average particle size: 65 ± 10 µm
Ligand: CaptureSelect™ IgA affinity ligand
Ligand coupling method: aldehyde coupling via NH2 residues of the ligand
Binding capacity: ~8 g/L resin
Elution conditions: 0.1 M glycine, pH 3.0
Flow characteristics: 200–400 cm/h
Formulation buffer: 20%(v/v) ethanol
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Column Type: Bulk Resin
Flow Rate: 200 to 400 cm⁄hr
Format: Bottle(s)
Immobilized Phase: Anti IgA
Particle Size: 65 micron
Product Line: CaptureSelect™
Product Size: 1 L
Support Matrix: Agarose based, aldehyde activated

Contents & storage

Store in refrigerator (2–8°C).

Documents

Manuals & protocols