CaptureSelect™ Human Growth Hormone Affinity Matrix
Thermo Scientific™

CaptureSelect™ Human Growth Hormone Affinity Matrix

Thermo Scientific™ CaptureSelect™ Human Growth Hormone (hGH) Affinity Matrix has been specifically designed for the purification of human growth hormone from recombinant sources.
Have Questions?
보기 방식 변경buttonViewtableView
카탈로그 번호수량
194316001010 mL
19431600055 mL
194316005050 mL
1943160250250 mL
194316001L1 L
194316005L5 L
카탈로그 번호 1943160010
제품 가격(KRW)
3,028,000
Each
수량:
10 mL
제품 가격(KRW)
3,028,000
Each

CaptureSelect Human Growth Hormone (hGH) Affinity Matrix has been specifically designed for the purification of human growth hormone from recombinant sources.

Features of this affinity matrix include:

  • Highly selective for recombinant hGH
  • Efficient purification of recombinant hGH with high yields and retained activity
  • Excellent scalability
  • Non-animal-derived

Recombinant hGH, also known as somatotropin or somatropin, is a peptide hormone that stimulates growth, cell reproduction, and regeneration in humans and other animals. The high selectivity and yields obtained using CaptureSelect Human Growth Hormone Affinity Matrix enable a robust and efficient purification process with excellent purity obtained in one step.

Free of animal components

CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12 to 15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).

Main characteristics:

  • Matrix: agarose-based, aldehyde-activated
  • Average particle size: 65 ± 10 μm
  • Ligand: CaptureSelect hGH affinity ligand
  • Ligand coupling method: aldehyde coupling via NH2 residues of the ligand
  • Binding capacity: ∼3 mg/mL resin
  • Elution conditions: 20 mM citric acid, pH 3.0
  • Flow characteristics: 50–200 cm/h (up to 2 bar)
  • Formulation buffer: 20%(v/v) ethanol
사양
결합 용량∼3 mg/mL
유속50 to 200 cm/hr. (Up to 2 bar)
리간드 유형HGH affinity ligand
수량10 mL
배송 조건Approved for shipment at Room Temperature and Wet Ice
정지상HGH
형태Agarose Bead
입자 크기65 ±10 μm
제품라인CaptureSelect
유형Affinity Matrix
Unit SizeEach
구성 및 보관
Store in refrigerator (2°C to 8°C).