CaptureSelect™ Human Growth Hormone Affinity Matrix
Thermo Scientific™

CaptureSelect™ Human Growth Hormone Affinity Matrix

Thermo Scientific™ CaptureSelect™ Human Growth Hormone (hGH) Affinity Matrix has been specifically designed for the purification of human growth hormone from recombinant sources.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
194316001L1 L
19431600055 mL
194316001010 mL
194316005050 mL
1943160250250 mL
194316005L5 L
Catalog number 194316001L
Price (CLP)
-
Quantity:
1 L

CaptureSelect Human Growth Hormone (hGH) Affinity Matrix has been specifically designed for the purification of human growth hormone from recombinant sources.

Features of this affinity matrix include:

  • Highly selective for recombinant hGH
  • Efficient purification of recombinant hGH with high yields and retained activity
  • Excellent scalability
  • Non-animal-derived

Recombinant hGH, also known as somatotropin or somatropin, is a peptide hormone that stimulates growth, cell reproduction, and regeneration in humans and other animals. The high selectivity and yields obtained using CaptureSelect Human Growth Hormone Affinity Matrix enable a robust and efficient purification process with excellent purity obtained in one step.

Free of animal components

CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12 to 15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).

Main characteristics:

  • Matrix: agarose-based, aldehyde-activated
  • Average particle size: 65 ± 10 μm
  • Ligand: CaptureSelect hGH affinity ligand
  • Ligand coupling method: aldehyde coupling via NH2 residues of the ligand
  • Binding capacity: ∼3 mg/mL resin
  • Elution conditions: 20 mM citric acid, pH 3.0
  • Flow characteristics: 50–200 cm/h (up to 2 bar)
  • Formulation buffer: 20%(v/v) ethanol
Specifications
Binding Capacity∼3 mg/mL
Flow Rate50 to 200 cm/hr. (Up to 2 bar)
Ligand TypeHGH affinity ligand
Quantity1 L
Shipping ConditionWet Ice
Stationary PhaseHGH
FormAgarose Bead
Particle Size65 ±10 μm
Product LineCaptureSelect
TypeAffinity Matrix
Unit SizeEach
Contents & Storage
Store in refrigerator (2°C to 8°C).