CaptureSelect™ Human Growth Hormone Affinity Matrix
Thermo Scientific™

CaptureSelect™ Human Growth Hormone Affinity Matrix

Thermo Scientific™ CaptureSelect™ Human Growth Hormone (hGH) Affinity Matrix has been specifically designed for the purification of human growth hormone from recombinant sources.
製品番号(カタログ番号)数量
194316005050 mL
19431600055 mL
194316001010 mL
1943160250250 mL
194316001L1 L
194316005L5 L
製品番号(カタログ番号) 1943160050
価格(JPY)
1,072,000
Each
数量:
50 mL

CaptureSelect Human Growth Hormone (hGH) Affinity Matrix has been specifically designed for the purification of human growth hormone from recombinant sources.

Features of this affinity matrix include:

  • Highly selective for recombinant hGH
  • Efficient purification of recombinant hGH with high yields and retained activity
  • Excellent scalability
  • Non-animal-derived

Recombinant hGH, also known as somatotropin or somatropin, is a peptide hormone that stimulates growth, cell reproduction, and regeneration in humans and other animals. The high selectivity and yields obtained using CaptureSelect Human Growth Hormone Affinity Matrix enable a robust and efficient purification process with excellent purity obtained in one step.

Free of animal components

CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12 to 15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).

Main characteristics:

  • Matrix: agarose-based, aldehyde-activated
  • Average particle size: 65 ± 10 μm
  • Ligand: CaptureSelect hGH affinity ligand
  • Ligand coupling method: aldehyde coupling via NH2 residues of the ligand
  • Binding capacity: ∼3 mg/mL resin
  • Elution conditions: 20 mM citric acid, pH 3.0
  • Flow characteristics: 50–200 cm/h (up to 2 bar)
  • Formulation buffer: 20%(v/v) ethanol
仕様
結合能∼3 mg/mL
流速50 to 200 cm/hr. (Up to 2 bar)
リガンドタイプHGH affinity ligand
数量50 mL
出荷条件Approved for shipment at Room Temperature and Wet Ice
固定相HGH
形状Agarose Bead
粒子径65 ±10 μm
製品ラインCaptureSelect
タイプAffinity Matrix
Unit SizeEach
組成および保存条件
Store in refrigerator (2°C to 8°C).