CaptureSelect™ Human Growth Hormone Affinity Matrix
Thermo Scientific™

CaptureSelect™ Human Growth Hormone Affinity Matrix

Thermo Scientific™ CaptureSelect™ Human Growth Hormone (hGH) Affinity Matrix has been specifically designed for the purification of human growth hormone from recombinant sources.
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoCantidad
194316005L5 L
19431600055 mL
194316001010 mL
194316005050 mL
1943160250250 mL
194316001L1 L
Número de catálogo 194316005L
Precio (MXN)
-
Cantidad:
5 L

CaptureSelect Human Growth Hormone (hGH) Affinity Matrix has been specifically designed for the purification of human growth hormone from recombinant sources.

Features of this affinity matrix include:

  • Highly selective for recombinant hGH
  • Efficient purification of recombinant hGH with high yields and retained activity
  • Excellent scalability
  • Non-animal-derived

Recombinant hGH, also known as somatotropin or somatropin, is a peptide hormone that stimulates growth, cell reproduction, and regeneration in humans and other animals. The high selectivity and yields obtained using CaptureSelect Human Growth Hormone Affinity Matrix enable a robust and efficient purification process with excellent purity obtained in one step.

Free of animal components

CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12 to 15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).

Main characteristics:

  • Matrix: agarose-based, aldehyde-activated
  • Average particle size: 65 μm
  • Ligand: CaptureSelect hGH affinity ligand
  • Ligand coupling method: aldehyde coupling via NH2 residues of the ligand
  • Binding capacity: ∼3 mg/mL resin
  • Elution conditions: 20 mM citric acid, pH 3.0
  • Flow characteristics: 50–200 cm/h (up to 2 bar)
  • Formulation buffer: 20%(v/v) ethanol
Especificaciones
Capacidad de unión∼3 mg/mL
Caudal50 to 200 cm/hr. (Up to 2 bar)
Tipo de ligandoHGH affinity ligand
Cantidad5 L
Condiciones de envíoWet Ice
Fase estacionariaHGH
Capacidad (métrico)∼3 mg/mL
FormularioResin Slurry
Tamaño de partículas65 μm
Línea de productosCaptureSelect
TipoAffinity Matrix
Unit SizeEach
Contenido y almacenamiento
• Operating Temperature: 2°C to 25°C
• Short-term Storage: 2°C to 25°C, avoid direct exposure to sunlight
• Long-term Storage: 2°C to 8°C, protected from light