CaptureSelect™ FSH Affinity Matrix
Thermo Scientific™

CaptureSelect™ FSH Affinity Matrix

Thermo Scientific™ CaptureSelect™ FSH Affinity Matrix has been specifically designed for the purification of follicle stimulating hormone (FSH) from recombinant sources.
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoCantidad
19431801010 mL
1943180055 mL
19431805050 mL
1943180250250 mL
19431801L1 L
19431805L5 L
Número de catálogo 194318010
Precio (MXN)
-
Cantidad:
10 mL

CaptureSelect FSH Affinity Matrix has been specifically designed for the purification of follicle stimulating hormone (FSH) from recombinant sources.

Free of animal components

CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12 to 15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).

For Research Use Only. Not for use in diagnostic procedures.
Especificaciones
Caudal200 to 400 cm/hr.
MatrizAgarose-based, aldehyde-activated
Cantidad10 mL
Fase estacionariaAnti-Follicle Stimulating Hormone
Capacidad (métrico)∼3 g FSH/L of matrix
FormularioSuspensión líquida
Tamaño de partículas65 μm
Línea de productosCaptureSelect
TipoAffinity Matrix
Unit SizeEach
Contenido y almacenamiento
• Operating Temperature: 2°C to 25°C
• Short-term Storage: 2°C to 25°C, avoid direct exposure to sunlight
• Long-term Storage: 2°C to 8°C, protected from light