CaptureSelect™ IgG-CH1 Affinity Matrix

Catalog number: 194320050

 Related applications: Antibody Purification | Chromatography | Protein Purification & Isolation

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The CaptureSelect™ IgG-CH1 affinity matrix recognizes the CH1 domain of human IgG antibodies, which enables purification of all subclasses (1, 2, 3 and 4) of human IgG and Fab fragments, independent of the light-chain isotype and source material. Due to its unique selectivity for the CH1 domain, no co-purification of free light-chain contaminants will occur. Since no cross-binding is observed for bovine IgG, the product can be used for the purification of IgGs and antibody fragments in the presence of fetal calf serum.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Column Type: Bulk Resin
Format: Bulk Media
Immobilized Phase: Anti IgG-CH1
Product Line: CaptureSelect™
Product Size: 50 mL

Contents & storage

Store at 2–8°C.

Documents

Manuals & protocols