Thermo Scientific™

CaptureSelect™ LC-lambda (rat) Affinity Matrix

Catalog number:  194324010

 Related applications:

Antibody Purification

| Chromatography

| Protein Purification & Isolation

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The CaptureSelect™ LC-lambda (rat) Affinity Matrix contains a 13 kDa llama antibody fragment recognizing the lambda light chain of Ig's from rat and guinea pig. The ligand is directed towards a unique domain on the constant part of the lambda light chain of immunoglobulins, which enables purification of rat and guinea pig immunoglobulins and fab fragments without cross reactivity towards bovine immunoglobulins. This makes the CaptureSelect™ LC-lambda (rat) Affinity Matrix a unique resin in terms of specificity. The ligand is coupled on a 70 µm aldehyde-activated agarose bead. This matrix can be used for the purification and isolation of lambda light chain-containing rat and guinea pig immunoglobulins and fab fragments from complex sources like cell culture supernatants, plasma, and serum in a single step. Due to its non-reactivity with bovine Ig, FCS can be applied in growth media without disturbing purification.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Column Type: Bulk Resin
Format: Bulk Media
Immobilized Phase: Anti LC-lambda
Product Line: CaptureSelect™
Product Size: 10 mL

Contents & storage

Store at 2–8°C.

Documents

Manuals & protocols