Search
Search

| Catalog Number | Quantity |
|---|---|
| 194341010 | 10 mL, 10 mL, 10 mL, 10 mL |
| 194341005 | 5 mL, 5 mL, 5 mL, 5 mL |
| 194341050 also known as 1943410050 | 50 mL, 50 mL, 50 mL, 50 mL |
| 1943410250 | 250 mL, 250 mL, 250 mL, 250 mL |
| 19434101L | 1 L, 1 L, 1 L, 1 L |
| 19434105L | 5 L, 5 L, 5 L, 5 L |
CaptureSelect hCG Affinity Matrix was specifically designed for the purification of human chorionic gonadotropin (hCG) from recombinant sources.
Some features of the CaptureSelect hCG Affinity Matrix include:
Human gonadotropins (hCG, LH, FSH, TSH) are glycoprotein hormones secreted by gonadotrope cells of the pituitary gland that are often produced as recombinant proteins. They have an identical alpha chain subunit that is recognized by the ligand. Consequently, the CaptureSelect hCG Affinity Matrix shows binding to all human gonadotropins. The high selectivity and yields obtained using CaptureSelect hCG Affinity Matrix enable a robust and efficient purification process for hCG with excellent purity obtained in one step.
Free of animal components
CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12 to 15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).
Main characteristics