Thermo Scientific™

CaptureSelect™ hCG Affinity Matrix

Catalog number: 194341050
Have Questions?
Thermo Scientific™

CaptureSelect™ hCG Affinity Matrix

Catalog number: 194341050
Have Questions?
Catalog Number
194341050
also known as 1943410050
Unit Size
50 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
5 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
10 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
250 mL
Price (USD)
Availability
-
Quantity
-
Catalog Number
Unit Size
1 L
Price (USD)
Availability
-
Quantity
-
Catalog Number
Unit Size
5 L
Price (USD)
Availability
-
Quantity
-
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
194341050
also known as 1943410050
50 mL
-
1943410055 mL
-
19434101010 mL
-
1943410250250 mL
--
19434101L1 L
--
19434105L5 L
--
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations

The CaptureSelect™ hCG Affinity Matrix was specifically designed for the purification of human chorionic gonadotropin (hCG) from recombinant sources.

Some features of the CaptureSelect hCG Affinity Matrix include:

  • Selective for hCG, with cross-binding to LH, FSH, and TSH
  • Enables efficient purification of recombinant gonadotropins with high yields and purity
  • Offers excellent scalability
  • Non–animal-derived

Human gonadotropins (hCG, LH, FSH, TSH) are glycoprotein hormones secreted by gonadotrope cells of the pituitary gland that are often produced as recombinant proteins. They have an identical alpha chain subunit that is recognized by the ligand. Consequently, the CaptureSelect hCG Affinity Matrix shows binding to all human gonadotropins. The high selectivity and yields obtained using CaptureSelect hCG Affinity Matrix enable a robust and efficient purification process for hCG with excellent purity obtained in one step.

Free of animal components

CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12 to 15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).

Main characteristics

  • Matrix: agarose-based, aldehyde-activated
  • Average particle size: 65 ± 10 μm
  • Ligand: CaptureSelect hCG affinity ligand
  • Ligand coupling method: aldehyde coupling via NH2 residues of the ligand
  • Binding capacity: approx. 4 mg/mL resin (determined with HCG)
  • Elution conditions: 20 mM citric acid, pH 3.0
  • Flow characteristics: 50–200 cm/h (up to 2 bar)
  • Formulation buffer: 20% (v/v) ethanol
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Shipping Condition
Room Temperature
Stationary Phase
Anti-gonadotropin
Product Line
CaptureSelect™
Type
Affinity Matrix
Quantity
50 mL
Form
Agarose Bead
Particle Size
65 ± 10 μm
Binding Capacity
4 mg/mL
Flow Rate
50 to 200 cm/hr. (Up to 2 bar)
Ligand Type
HCG affinity ligand
Matrix
Agarose-based, aldehyde-activated

Contents & Storage

Store in refrigerator (2°C to 8°C).

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text