Ammonium hexafluorotitanate(IV), 99.99%, (trace metal basis)
Ammonium hexafluorotitanate(IV), 99.99%, (trace metal basis)
Ammonium hexafluorotitanate(IV), 99.99%, (trace metal basis)
Thermo Scientific Chemicals

Ammonium hexafluorotitanate(IV), 99.99%, (trace metal basis)

CAS: 16962-40-6 | H8F6N2Ti | 197.97 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
Catalog number 194350100
Price (USD)
-
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS16962-40-6
IUPAC Nametitanium(4+) diammonium hexafluoride
Molecular FormulaF6H8N2Ti
InChI KeyNMGYKLMMQCTUGI-UHFFFAOYSA-J
SMILES[NH4+].[NH4+].[F-].[F-].[F-].[F-].[F-].[F-].[Ti+4]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals and/or chunks
Total trace metal impurities=<0.01 %
Appearance (Color)White
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for ultra trace analysis
RUO – Research Use Only