CaptureSelect™ IgE Affinity Matrix
Thermo Scientific™

CaptureSelect™ IgE Affinity Matrix

Thermo Scientific™ CaptureSelect™ IgE Affinity Matrix has been designed specifically for the purification of human IgE (immunoglobuline E) from recombinant and plasma sources.
Have Questions?
Alterar visualizaçãobuttonViewtableView
Número do catálogoQuantity
194354201L1 L
29435420055 mL
294354201010 mL
294354205050 mL
1943542250250 mL
194354205L5 L
Número do catálogo 194354201L
Preço (BRL)
-
Quantity:
1 L

CaptureSelect IgE Affinity Matrix has been designed specifically for the purification of human IgE (immunoglobuline E) from recombinant and plasma sources.

Features of this affinity matrix include:

  • Purification of human IgE, binding different recombinant IgE forms
  • Efficient removal of host cell proteins (HCP) and IgE aggregates
  • Excellent scalability
  • Non–animal-derived

Free of animal components

CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12 to 15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).

Main characteristics

  • Matrix: agarose-based, epoxide-activated
  • Average particle size: 65 μm
  • Ligand: CaptureSelect IgE affinity ligand
  • Ligand coupling method: aldehyde coupling
  • Binding capacity: >5 mg/mL resin
  • Elution conditions: 50 mM sodium citrate, 150 mM NaCl, pH 3.5
  • Flow characteristics: 50–200 cm/h (up to 2 bar)
  • Formulation buffer: 20% (v/v) ethanol
Especificações
Binding Capacity>5 mg/mL resin
Flow Rate50 to 200 cm/hr. (Up to 2 bar)
Ligand TypeCaptureSelect IgE Affinity Ligand
MatrixAgarose
Max. Operating Pressure2 bar
Quantity1 L
Stationary PhaseIgE
Capacity (Metric)>5 mg/mL resin
FormResin Slurry
Particle Size65 μm
Product LineCaptureSelect
TypeAffinity Matrix
Unit SizeEach
Conteúdo e armazenamento
• Operating Temperature: 2°C to 25°C
• Short-term Storage: 2°C to 25°C, avoid direct exposure to sunlight
• Long-term Storage: 2°C to 8°C, protected from light