Thermo Scientific™

CaptureSelect™ IgE Affinity Matrix

Catalog number: 1943542250
Have Questions?
Thermo Scientific™

CaptureSelect™ IgE Affinity Matrix

Catalog number: 1943542250
Have Questions?
Catalog Number
1943542250
Unit Size
250 mL
Price (USD)
Availability
-
Quantity
-
Catalog Number
Unit Size
5 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
10 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
50 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
1000 mL
Price (USD)
Availability
-
Quantity
-
Catalog Number
Unit Size
5000 mL
Price (USD)
Availability
-
Quantity
-
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
1943542250250 mL
--
29435420055 mL
-
294354201010 mL
-
294354205050 mL
-
194354201L1000 mL
--
194354205L5000 mL
--
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations

Thermo Scientific CaptureSelect IgE Affinity Matrix has been designed specifically for the purification of human IgE (immunoglobuline E) from recombinant and plasma sources.

Features of this affinity matrix include:

  • Purification of human IgE, binding different recombinant IgE forms
  • Efficient removal of host cell proteins (HCP) and IgE aggregates
  • Excellent scalability
  • Non–animal-derived

Free of animal components

CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12 to 15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).

Main characteristics

  • Matrix: agarose-based, epoxide-activated
  • Average particle size: 65 ± 10 μm
  • Ligand: CaptureSelect IgE affinity ligand
  • Ligand coupling method: aldehyde coupling
  • Binding capacity: >5 mg/mL resin
  • Elution conditions: 50 mM sodium citrate, 150 mM NaCl, pH 3.5
  • Flow characteristics: 50–200 cm/h (up to 2 bar)
  • Formulation buffer: 20% (v/v) ethanol

Specifications

Binding Capacity
>5 mg/mL resin
Flow Rate
50 to 200 cm/hr. (Up to 2 bar)
Matrix
Agarose-based, epoxide-activated
Particle Size
65 ± 10 μm
Form
Resin Slurry
Product Line
CaptureSelect™
Quantity
250 mL
Type
Affinity Matrix
Ligand Type
IgE affinity ligand
Stationary Phase
IgE

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text