Thermo Scientific™

CaptureSelect™ TSH Affinity Matrix

Catalog number: 194356205L
Have Questions?
Thermo Scientific™

CaptureSelect™ TSH Affinity Matrix

Catalog number: 194356205L
Have Questions?
Catalog Number
194356205L
Unit Size
5 L
Price (USD)
Availability
-
Quantity
-
Catalog Number
Unit Size
5 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
10 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
50 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
250 mL
Price (USD)
Availability
-
Quantity
-
Catalog Number
Unit Size
1 L
Price (USD)
Availability
-
Quantity
-
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
194356205L5 L
--
29435620055 mL
-
294356201010 mL
-
294356205050 mL
-
1943562250250 mL
--
194356201L1 L
--
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations

CaptureSelect TSH Affinity Matrix was specifically designed for the purification of thyroid-stimulating hormone (TSH) from recombinant sources.

Some features of CaptureSelect TSH Affinity Matrix include:

  • Highly selective for thyroid-stimulating hormone with no affinity for the other gonadotropins (FSH, LH, and HCG)
  • Enables efficient purification of recombinant TSH with high yields and retained activity
  • Offers excellent scalability
  • Non–animal-derived

The high selectivity and yields obtained using CaptureSelect TSH Affinity Matrix enable a robust and efficient purification process with excellent purity obtained in one step.

Free of animal components

CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12 to 15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).

Main characteristics:

  • Matrix: agarose bead
  • Average particle size: 65 μm
  • Ligand: CaptureSelect TSH affinity ligand
  • Ligand coupling method: epoxide
  • Binding capacity: >5 mg/mL of resin
  • Elution conditions: 0.1 M citrate buffer, pH 3.0
  • Flow characteristics: 50–200 cm/h (up to 2 bar)
  • Formulation buffer: 20% (v/v) ethanol

Specifications

Binding Capacity
>5 mg/mL
Flow Rate
50 to 200 cm/hr. (Up to 2 bar)
Matrix
Agarose-based
Shipping Condition
Wet Ice
Particle Size
65 μm
Form
Agarose Bead
Product Line
CaptureSelect™
Quantity
5 L
Type
Affinity Matrix
Ligand Type
TSH affinity ligand
Stationary Phase
Thyroid Stimulating Hormone (TSH)

Contents & Storage

Store in refrigerator (2°C to 8°C).

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text