Yttrium(III) oxide, 99.999%, (trace metal basis)
Yttrium(III) oxide, 99.999%, (trace metal basis)
Yttrium(III) oxide, 99.999%, (trace metal basis)
Thermo Scientific Chemicals

Yttrium(III) oxide, 99.999%, (trace metal basis)

CAS: 1314-36-9 | O3Y2 | 225.81 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 g
50 g
Packaging:
Glass bottle
Catalog number 194480020
Price (USD)
-
Quantity:
2 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS1314-36-9
IUPAC Namediyttrium(3+) trioxidandiide
Molecular FormulaO3Y2
InChI KeyRUDFQVOCFDJEEF-UHFFFAOYSA-N
SMILES[O--].[O--].[O--].[Y+3].[Y+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay99.999 % (Y2O3) On TREO base
Appearance (Color)White
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for ultra trace analysis
RUO – Research Use Only