Titanium(IV) isopropoxide, 98+%
Titanium(IV) isopropoxide, 98+%
Titanium(IV) isopropoxide, 98+%
Thermo Scientific Chemicals

Titanium(IV) isopropoxide, 98+%

CAS: 546-68-9 | C12H28O4Ti | 284.26 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mL
250 mL
1 L
2.5 L
Packaging:
Glass bottle
Catalog number 194700010
Price (USD)
145.00
Each
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
145.00
Each
Chemical Identifiers
CAS546-68-9
IUPAC Nametitanium(4+) tetrakis(propan-2-olate)
Molecular FormulaC12H28O4Ti
InChI KeyVXUYXOFXAQZZMF-UHFFFAOYSA-N
SMILES[Ti+4].CC(C)[O-].CC(C)[O-].CC(C)[O-].CC(C)[O-]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
Color scale=<100 APHA
AppearanceMay darken on storage
Infrared spectrumConforms
Melting point>=15°C
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis acid
RUO – Research Use Only