Lithium perchlorate, 99+%, for analysis
Lithium perchlorate, 99+%, for analysis
Lithium perchlorate, 99+%, for analysis
Lithium perchlorate, 99+%, for analysis
Thermo Scientific Chemicals

Lithium perchlorate, 99+%, for analysis

CAS: 7791-03-9 | ClLiO4 | 106.39 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
500 g
Packaging:
View all
Glass Bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 194710050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS7791-03-9
IUPAC Namelithium(1+) trihydrate perchlorate
Molecular FormulaClH6LiO7
InChI KeyJAWYRNYHJJDXHX-UHFFFAOYSA-M
SMILES[Li+].O.O.O.[O-][Cl](=O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Crystalline or granular powder or crystals
Titration after Ion exchange99.0 to 101.0 %
Water=<1.0 % (K.F.)
Insoluble matter=<0.005 % (in water)
View more

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Oxidation reagent for analysis
RUO – Research Use Only